Få mere at vide
6-[(3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethenyl] Nicotinic Acid Methyl Ester, TRC
Artikelnummer. 30427768
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427768 10 mg
30408973 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30427768 Leverandør Toronto Research Chemicals Leverandørnr. TRCP27178010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-[(3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethenyl] Nicotinic Acid Methyl Ester
CAS 153559-44-5
Molekylær formel C24H29NO2
Mængde 10 mg
Synonym 6-[1-(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)ethenyl]-3-pyridinecarboxylic Acid Methyl Ester
InChI formel InChI=1S/C24H29NO2/c1-15-12-19-20(24(5,6)11-10-23(19,3)4)13-18(15)16(2)21-9-8-17(14-25-21)22(26)27-7/h8-9,12-14H,2,10-11H2,1,3-7H3
SMIL COC(=O)c1ccc(nc1)C(=C)c2cc3c(cc2C)C(C)(C)CCC3(C)C
IUPAC navn methyl 6-[1-(3,5,5,8,8-pentamethyl-6,7-dihydronaphthalen-2-yl)ethenyl]pyridine-3-carboxylate
Molekylvægt (g/mol) 363.49
Formel vægt 363.2198