missing translation for 'onlineSavingsMsg'
Få mere at vide

6-[(3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethenyl] Nicotinic Acid Methyl Ester, TRC

Artikelnummer. 30408973
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30408973 5 mg
30427768 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30408973 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2717805MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 6-[(3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethenyl] Nicotinic Acid Methyl Ester
CAS 153559-44-5
Molekylær formel C24H29NO2
Mængde 5 mg
Synonym 6-[1-(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)ethenyl]-3-pyridinecarboxylic Acid Methyl Ester
InChI formel InChI=1S/C24H29NO2/c1-15-12-19-20(24(5,6)11-10-23(19,3)4)13-18(15)16(2)21-9-8-17(14-25-21)22(26)27-7/h8-9,12-14H,2,10-11H2,1,3-7H3
SMIL COC(=O)c1ccc(nc1)C(=C)c2cc3c(cc2C)C(C)(C)CCC3(C)C
IUPAC navn methyl 6-[1-(3,5,5,8,8-pentamethyl-6,7-dihydronaphthalen-2-yl)ethenyl]pyridine-3-carboxylate
Molekylvægt (g/mol) 363.49
Formel vægt 363.2198
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.