Få mere at vide
6-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine Hydrochloride, Mikromol™
Beskrivelse
6-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 6-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine Hydrochloride |
| CAS | 2985091-68-5 |
| Analyt- eller komponentnavne | 6-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C14 H14 Cl N S . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Ticlopidine Hydrochloride Imp. F (EP), Ticlopidine Imp. F (EP), 6-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine Hydrochloride, Ticlopidine Hydrochloride Imp. F (EP) as Hydrochloride, Ticlopidine Hydrochloride Impurity F as Hydrochloride, Ticlopidine Impurity F as Hydrochloride |
| InChI formel | InChI=1S/C17H20N2S.ClH/c1-13(12-18(2)3)19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H |
| SMIL | Cl.Clc1ccccc1CN2CCc3ccsc3C2 |
| IUPAC navn | 6-[(2-chlorophenyl)methyl]-5,7-dihydro-4H-thieno[2,3-c]pyridine;hydrochloride |
| Vis mere |