Få mere at vide
6-(2-Aminoethylamino)-7-chloro-1-cyclopropyl-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid Hydrochloride, Mikromol™
Artikelnummer. 30461074
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30461074 4 x 25 mg
30052130 100 mg
30050302 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30461074 Leverandør Mikromol Leverandørnr. MM0018.1000254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-(2-Aminoethylamino)-7-chloro-1-cyclopropyl-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-(2-Aminoethylamino)-7-chloro-1-cyclopropyl-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid Hydrochloride
CAS 528851-30-1
Analyt- eller komponentnavne 6-(2-Aminoethylamino)-7-chloro-1-cyclopropyl-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C15 H16 Cl N3 O3 . Cl H
Certificeringer/compliance ISO 9001
Synonym 6-(2-Aminoethylamino)-7-chloro-1-cyclopropyl-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid Hydrochloride
InChI formel InChI=1S/C15H16ClN3O3.ClH/c16-11-6-13-9(5-12(11)18-4-3-17)14(20)10(15(21)22)7-19(13)8-1-2-8;/h5-8,18H,1-4,17H2,(H,21,22);1H
Løsningstype Neat
SMIL Cl.NCCNc1cc2C(=O)C(=CN(C3CC3)c2cc1Cl)C(=O)O
IUPAC navn 6-(2-aminoethylamino)-7-chloro-1-cyclopropyl-4-oxoquinoline-3-carboxylic acid;hydrochloride
Molekylvægt (g/mol) 358.22
Mængde 4 x 25 mg
Formel vægt 357.0647 g/mol