Få mere at vide
6-[(1E, 3E)-4-(4-Methoxy-2,3,6-trimethylphenyl-d3)-2-methyl-1,3-butadien-1-yl]-4-methyl-2H-pyran-2-one, TRC
Artikelnummer. 30476735
Skift visning
Mængde:
2 mg
Artikelnummer. Mængde
30476735 2 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476735 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2715022MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-[(1E','3E)-4-(4-Methoxy-2,3,6-trimethylphenyl-d3)-2-methyl-
CAS 1185236-53-6
Molekylær formel C21 2H3 H21 O3
Mængde 2 mg
InChI formel InChI=1S/C21H24O3/c1-13(9-18-10-14(2)11-21(22)24-18)7-8-19-15(3)12-20(23-6)17(5)16(19)4/h7-12H,1-6H3/b8-7+,13-9+/i6D3
SMIL [2H]C([2H])([2H])Oc1cc(C)c(\C=C\C(=C\C2=CC(=CC(=O)O2)C)\C)c(C)c1C
IUPAC navn 4-methyl-6-[(1E,3E)-2-methyl-4-[2,3,6-trimethyl-4-(trideuteriomethoxy)phenyl]buta-1,3-dienyl]pyran-2-one
Molekylvægt (g/mol) 327.43
Formel vægt 327.1914