Få mere at vide
6,13-Pentacenedione-d12, TRC
Artikelnummer. 30416459
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30416459 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30416459 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2377821MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6,13-Pentacenedione-d12
CAS 68234-48-0
Molekylær formel C22D12O2
Mængde 1 mg
Synonym 6,13-Pentacenequinone-d12
InChI formel InChI=1S/C22H12O2/c23-21-17-9-13-5-1-2-6-14(13)10-18(17)22(24)20-12-16-8-4-3-7-15(16)11-19(20)21/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D
SMIL O=C1C2=C([2H])C3=C([2H])C([2H])=C([2H])C([2H])=C3C([2H])=C2C(C4=C([2H])C5=C([2H])C([2H])=C([2H])C([2H])=C5C([2H])=C41)=O
IUPAC navn pentacene-6,13-dione-d12
Molekylvægt (g/mol) 320.4
Formel vægt 320.16