missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 6-(1-Piperidinyl)pyridine-3-carboxaldehyde, 98%
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | 42°C to 44°C |
Quantity | 1 g |
Formula Weight | 190.25 |
Percent Purity | 98% |
Chemical Name or Material | 6-(1-Piperidinyl)pyridine-3-carboxaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'