Få mere at vide
6-(1-Fluoroethyl)-1,3,5-triazine-2,4-diamine, TRC
Artikelnummer. 30179857
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179857 500 mg
30180214 250 mg
30179816 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179857 Leverandør Toronto Research Chemicals Leverandørnr. TRCF532080500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1637285-20-1
Molekylær formel C5 H8 F N5
Molekylvægt (g/mol) 157.15
Synonym 1,3,5-Triazine-2,4-diamine, 6-(1-fluoroethyl)-
IUPAC navn 6-(1-fluoroethyl)-1,3,5-triazine-2,4-diamine
SMIL CC(F)c1nc(N)nc(N)n1
Tekniske data
Kemisk navn eller materiale Indaziflam-desindenyl
Mængde 500 mg
InChI formel InChI=1S/C5H8FN5/c1-2(6)3-9-4(7)11-5(8)10-3/h2H,1H3,(H4,7,8,9,10,11)
Formel vægt 157.0764