Få mere at vide
5R-Hydroxy-L-lysine Dihydrochloride Salt, TRC
Artikelnummer. 30136743
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136743 10 mg
30012357 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136743 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94442010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 172213-74-0
Molekylær formel C6H14N2O3 . 2(HCl)
Molekylvægt (g/mol) 235.11
Synonym (5R)-5-Hydroxy-L-lysine Dihydrochloride, (2S,5R)-5-Hydroxylysine Dihydrochloride, 5-Hydroxylysine Dihydrochloride, Hydroxylysine Dihydrochloride
SMIL NC[C@H](O)CC[C@H](N)C(=O)O
Tekniske data
Kemisk navn eller materiale 5R-Hydroxy-L-lysine Dihydrochloride Salt
InChI formel InChI=1S/C6H14N2O3.2ClH/c7-3-4(9)1-2-5(8)6(10)11;;/h4-5,9H,1-3,7-8H2,(H,10,11);2*1H