Få mere at vide
(5R)-5-(Chloromethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone, Mikromol™
Artikelnummer. 30050902
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050902 100 mg
30049395 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050902 Leverandør Mikromol Leverandørnr. MM3300.08
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(5R)-5-(Chloromethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (5R)-5-(Chloromethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone
CAS 496031-57-3
Analyt- eller komponentnavne (5R)-5-(Chloromethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone
Forsendelsestilstand Room Temperature
Molekylær formel C14 H16 Cl F N2 O3
Certificeringer/compliance ISO 17025
Synonym 9-Allylidene-2-(trifluoromethyl)thioxanthene 10-Oxide, 2-Oxazolidinone, 5-(chloromethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-, (5R)-, (5R)-5-(Chloromethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone
InChI formel InChI=1S/C14H16ClFN2O3/c15-8-11-9-18(14(19)21-11)10-1-2-13(12(16)7-10)17-3-5-20-6-4-17/h1-2,7,11H,3-6,8-9H2/t11-/m0/s1
SMIL Fc1cc(ccc1N2CCOCC2)N3C[C@H](CCl)OC3=O
IUPAC navn (5R)-5-(chloromethyl)-3-(3-fluoro-4-morpholin-4-ylphenyl)-1,3-oxazolidin-2-one
Molekylvægt (g/mol) 314.74
Mængde 100 mg
Formel vægt 314.741 g/mol