Få mere at vide
5beta-Pregnan-3alpha-ol-20-one-17alpha,21,21,21-d4, CDN
Artikelnummer. 30315156
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315156 0.01 g
30312257 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315156 Leverandør CDN Leverandørnr. CDND65790.01G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5 beta-Pregnan-3 alpha-ol-20-one-d4
CAS 96737-93-8
Molekylær formel C21 D4 H30 O2
Mængde 0.01 g
Synonym Pregnan-20-one-17,21,21,21-d4, 3-hydroxy-, (3α,5β)- (9 CI), (3α,5β)-3-Hydroxypregnan-20-one-17,21,21,21-d4, 3α-Hydroxy-5β-pregnan-20-one-17,21,21,21-D4, Pregnanolone-17,21,21,21-d4, 3α,5β-Pregnanolone-17,21,21,21-d4, 3α,5β-Tetrahydroprogesterone-17,21,21,21-d4, 5β-Pregnan-3α-ol-20-one-17α,21,21,21-d4, 5β-Pregnan-3α-ol-20-one-d4
InChI formel InChI=1 S/C21H34O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h14-19,23 H,4-12H2,1-3H3/t14-,15-,16+,17-,18+,19+,20+,21-/m1/s1/i1D3,17 D
SMIL [2 H]C([2 H])([2 H])C(=O)[C@@]1([2 H])CC[C@H]2 [C@@H]3 CC[C@@H]4 C[C@H](O)CC[C@]4(C)[C@H]3 CC[C@]12 C
IUPAC navn 2,2,2-trideuterio-1-[(3 R,5 R,8 R,9 S,10 S,13 S,14 S,17 S)-17-deuterio-3-hydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-17-yl]ethanone
Molekylvægt (g/mol) 322.518
Formel vægt 322.281 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity