Få mere at vide
5beta-Pregnan-3alpha,21-diol-20-one-17alpha,21,21-d3, TRC
Artikelnummer. 30419203
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30419203 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419203 Leverandør Toronto Research Chemicals Leverandørnr. TRCT2932715MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5beta-Pregnan-3alpha,21-diol-20-one-17alpha,21,21-d3
CAS 72205-58-4
Molekylær formel C21 D3 H31 O3
Mængde 5 mg
Synonym Pregnan-20-one-17,21,21-d3','3,21-dihydroxy-','(3α,5β)- (9CI), 5β-Pregnan-3α,21-diol-20-one-17α,21,21-d3, (3α,5β)-3,21-Dihydroxypregnan-20-one-17α,21,21-d3, 3α,21-Dihydroxy-5β-pregnan-20-one-17,21,21-D3, Tetrahydro-11-deoxycorticosterone-17,21,21-d3
InChI formel InChI=1S/C21H34O3/c1-20-9-7-14(23)11-13(20)3-4-15-16-5-6-18(19(24)12-22)21(16,2)10-8-17(15)20/h13-18,22-23H,3-12H2,1-2H3/t13-,14-,15+,16+,17+,18-,20+,21+/m1/s1/i12D2,18D
SMIL [2H]C([2H])(O)C(=O)[C@@]1([2H])CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC navn 2,2-dideuterio-1-[(3R,5R,8R,9S,10S,13S,14S,17S)-17-deuterio-3-hydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-17-yl]-2-hydroxyethanone
Molekylvægt (g/mol) 337.5114
Formel vægt 337.2696