Få mere at vide
5beta-Pregnan-3alpha,17alpha,21-triol-11,20-dione-9,12,12,21,21-d5, CDN
Artikelnummer. 30318941
Skift visning
Mængde:
0.005 g
Artikelnummer. Mængde
30318941 0.005 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318941 Leverandør CDN Leverandørnr. CDND65900.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tetrahydro Cortisone-d5
CAS 1485501-48-1
Molekylær formel C21 D5 H27 O5
Mængde 0.005 g
Synonym (3α,5β)-3,17,21-Trihydroxypregnane-11,20-dione-d5, 3α,17,21-Trihydroxy-5β-pregnan-11,20-dione-d4, 5β-Pregnan-3α,17α,21-triol- 11,20-dione-d5, 5β-Tetrahydrocortisone-d5, Ba 2681-d5, 5 beta-Pregnan-3 alpha,17 alpha,21-triol-11,20-dione-9,12,12,21,21-d5, 5 beta-Pregnan-3 alpha,17 alpha,21-triol-11,20-dione-9,12,12,21,21-d5, 5 beta-Pregnan-3 alpha,17 alpha,21-triol-11,20-dione-9,12,12,21,21-d5, 5 beta-Pregnan-3 alpha,17 alpha,21-triol-11,20-dione-9,12,12,21,21-d5, 5 beta-Pregnan-3 alpha,17 alpha,21-triol-11,20-dione-9,12,12,21,21-d5, 5 beta-Pregnan-3 alpha,17 alpha,21-triol-11,20-dione-9,12,12,21,21-d5, 5 beta-Pregnan-3 alpha,17 alpha,21-triol-11,20-dione-9,12,12,21,21-d5
InChI formel InChI=1 S/C21H32O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h12-15,18,22-23,26 H,3-11H2,1-2H3/t12-,13-,14+,15+,18-,19+,20+,21+/m1/s1/i10D2,11D2,18 D
SMIL [2 H]C([2 H])(O)C(=O)[C@@]1(O)CC[C@H]2 [C@@H]3 CC[C@@H]4 C[C@H](O)CC[C@]4(C)[C@@]3([2 H])C(=O)C([2 H])([2 H])[C@]12 C
IUPAC navn (3 R,5 R,8 S,9 S,10 S,13 S,14 S,17 R)-9,12,12-trideuterio-17-(2,2-dideuterio-2-hydroxyacetyl)-3,17-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,14,15,16-decahydro-1 H-cyclopenta[a]phenanthren-11-one
Molekylvægt (g/mol) 369.51
Formel vægt 369.2564 g/mol
Procent renhed 98 atom % D, min 96% Chemical Purity