Få mere at vide
5Beta-Androsterone Beta-D-Glucuronide, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Etiocholanolone glucuronide |
| CAS | 3602-09-3 |
| Molekylær formel | C25H38O8 |
| Mængde | 10 mg |
| Synonym | 5β-Androsterone β-D-Glucuronide, β-D-Glucopyranosiduronic acid, (3α,5β)-17-oxoandrostan-3-yl, Glucopyranosiduronic acid, 17-oxo-5β-androstan-3α-yl, β-D- (6CI,8CI), (3α,5β)-17-Oxoandrostan-3-yl β-D-glucopyranosiduronic acid, 3α-Hydroxy-5β-androstan-17-one glucuronide, 5β-Androstan-3α-ol-17-one glucuronide, Androstan-3-ol-17-one 3-glucuronide, Etiocholan-3α-ol-17-one 3-glucuronide, Etiocholan-3α-ol-17-one glucuronide, Etiocholanolone glucuronide, Etiocholanolone glucuronoside, NSC 113100 |
| SMIL | C[C@]12CC[C@H](C[C@H]1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CCC4=O)O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)O |
| IUPAC navn | (2S,3S,4S,5R,6R)-6-[[(3R,5R,8R,9S,10S,13S,14S)-10,13-dimethyl-17-oxo-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-3-yl]oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid |
| Molekylvægt (g/mol) | 466.5 |
| Formel vægt | 466.5644 |