Få mere at vide
5beta-Androstan-3alpha,17beta-diol-2,2,3,4,4-d5, CDN
Artikelnummer. 30323247
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323247 0.01 g
30320894 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323247 Leverandør CDN Leverandørnr. CDND74400.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale d5-5 b-Androstane-3 A,17 b-diol
CAS 1224710-37-5
Molekylær formel C19 2H5 H27 O2
Mængde 0.01 g
SMIL [2 H]C1([2 H])C[C@]2(C)[C@H]3 CC[C@]4(C)[C@@H](O)CC[C@H]4 [C@@H]3 CC[C@@H]2 C([2 H])([2 H])[C@]1([2 H])O
IUPAC navn (3 R,5 R,8 R,9 S,10 S,13 S,14 S,17 S)-2,2,3,4,4-pentadeuterio-10,13-dimethyl-1,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthrene-3,17-diol
Molekylvægt (g/mol) 297.4
Formel vægt 297.487 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity