missing translation for 'onlineSavingsMsg'
Få mere at vide

5b-Androstane-3a,17b-diol Diacetate, TRC

Product Code. 30348123
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30348123 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348123 Supplier Toronto Research Chemicals Supplier No. TRCA63771025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 5beta-Androstane-3alpha,17beta-diol Diacetate
CAS 4062-43-5
Molekylær formel C23H36O4
Mængde 25 mg
Synonym (3α,5β,17β)-Androstane-3,17-diol, 3,17-diacetate, (3α,5β,17β)- Androstane-3,17-diol, Diacetate
InChI formel InChI=1S/C23H36O4/c1-14(24)26-17-9-11-22(3)16(13-17)5-6-18-19-7-8-21(27-15(2)25)23(19,4)12-10-20(18)22/h16-21H,5-13H2,1-4H3/t16-,17-,18+,19+,20+,21+,22+,23+/m1/s1
SMIL [H][C@@]12C[C@H](OC(C)=O)CC[C@]1(C)[C@]3([H])[C@@]([C@@](CC[C@@H]4OC(C)=O)([H])[C@]4(C)CC3)([H])CC2
IUPAC navn [(3R,5R,8R,9S,10S,13S,14S,17S)-17-acetyloxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
Molekylvægt (g/mol) 376.53
Formel vægt 376.2614
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.