Få mere at vide
5alpha-Pregnan-3alpha-ol-20-one-17alpha,21,21,21-d4, TRC
Artikelnummer. 30315331
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30315331 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315331 Leverandør Toronto Research Chemicals Leverandørnr. TRCA54710110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3alpha-Hydroxy-5alpha-pregnan-20-one-17,21,21,21-D4
CAS 203805-85-0
Molekylær formel C21 H30 D4 O2
Mængde 10 mg
Synonym Pregnan-20-one-17,21,21,21-d4, 3-hydroxy-, (3α,5α)- (9CI, ACI), (3α,5α)-3-Hydroxypregnan-20-one-17,21,21,21-d4 (ACI), (3α)-Allopregnanolone-d4, Brexanolone-17,21,21,21-d4, 3α-Hydroxy-5α-pregnan-20-one-17,21,21,21-D4, Allopregnanolone-17,21,21,21-d4, 3α,5α-Pregnanolone-17,21,21,21-d4, 3α,5α-Tetrahydroprogesterone-17,21,21,21-d4, 5α-Pregnan-3α-ol-20-one-17α,21,21,21-d4, 5alpha-Pregnan-3alpha-ol-20-one-17alpha,21,21,21-d4, Brexanolone-D4
InChI formel InChI=1S/C21H34O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h14-19,23H,4-12H2,1-3H3/t14-,15+,16-,17+,18-,19-,20-,21+/m0/s1/i1D3,17D
SMIL C[C@@]12[C@@]3([C@]([C@]4([C@](C)(CC3)[C@@](C(C([2H])([2H])[2H])=O)(CC4)[2H])[H])(CC[C@]1(C[C@H](O)CC2)[H])[H])[H]
IUPAC navn 2,2,2-trideuterio-1-[(3R,5S,8R,9S,10S,13S,14S,17S)-17-deuterio-3-hydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-17-yl]ethanone
Molekylvægt (g/mol) 322.51811
Formel vægt 322.281