Få mere at vide
5Alpha-Dihydrotestosterone (2,2,4,4-D4, 98%) 95% Chemical Purity, TRC
Artikelnummer. 30371874
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30371874 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371874 Leverandør Toronto Research Chemicals Leverandørnr. TRCD45950810MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5alpha-Dihydrotestosterone 2,2,4,4-D4
CAS 5295-66-9
Molekylær formel C19 D4 H26 O2
Mængde 10 mg
Synonym Androstan-3-one-2,2,4,4-d4, 17-hydroxy-, (5α,17β)- (9CI, ACI), (5α,17β)-17-Hydroxyandrostan-3-one-2,2,4,4-d4 (ACI), 5α-Androstan-3-one-2,2,4,4-d4, 17β-hydroxy- (7CI, 8CI), Androstanolone-d4, Androstanolone-2,2,4,4-d4, Androstanolone-2,2,4,4-D4, 5α-Dihydrotestosterone-2,2,4,4-d4, 5α-Androstan-17β-ol-3-one-2,2,4,4-d4
InChI formel InChI=1S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12,14-17,21H,3-11H2,1-2H3/t12-,14-,15-,16-,17-,18-,19-/m0/s1/i7D2,11D2
SMIL [2H]C1([2H])C[C@]2(C)[C@H]3CC[C@]4(C)[C@@H](O)CC[C@H]4[C@@H]3CC[C@H]2C([2H])([2H])C1=O
IUPAC navn (5S,8R,9S,10S,13S,14S,17S)-2,2,4,4-tetradeuterio-17-hydroxy-10,13-dimethyl-1,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one
Molekylvægt (g/mol) 294.465
Formel vægt 294.25