Få mere at vide
5alpha-Dihydrocortisol, TRC
Artikelnummer. 30138141
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138141 5 mg
30134174 10 mg
30008756 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138141 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4488605MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 516-41-6
Molekylær formel C21 H32 O5
Molekylvægt (g/mol) 364.48
Synonym Pregnane-3,20-dione, 11,17,21-trihydroxy-, (5α,11β)-, 5α-Pregnane-3,20-dione, 11β,17,21-trihydroxy- (7CI,8CI), (5α,11β)-11,17,21-Trihydroxypregnane-3,20-dione, 11β,17,21-Trihydroxy-5α-pregnane-3,20-dione, 11β,17α,21-Trihydroxy-5α-pregnane-3,20-dione, 5α-Dihydrocortisol, 5α-Pregnan-11β,17α,21-triol-3,20-dione, Allodihydrocortisol, Allodihydrohydrocortisone, Allopregnane-11β,17α,21-triol-3,20-dione, Hydrallostane
IUPAC navn (5S,8S,9S,10S,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,4,5,6,7,8,9,11,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-3-one
SMIL C[C@]12CCC(=O)C[C@@H]1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23
Tekniske data
Kemisk navn eller materiale 5-alpha-Dihydrocortisol
Mængde 5 mg
InChI formel InChI=1S/C21H32O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h12,14-16,18,22,24,26H,3-11H2,1-2H3/t12-,14-,15-,16-,18+,19-,20-,21-/m0/s1
Formel vægt 364.225