Få mere at vide
5Alpha-Cholestane-2,2,3,3,4,4-d6, TRC
Artikelnummer. 30355201
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30355201 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30355201 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4317035MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5a-Cholestane-2,2,3,3,4,4-d6
CAS 358731-18-7
Molekylær formel C27H42D6
Mængde 5 mg
Synonym Cholestane-2,2,3,3,4,4-d6, (5α)- (9CI), 5α-Cholestane-2,2,3,3,4,4-D6, 5α-Cholestane-2,2,3,3,4,4-d6, 5α-Cholestane-d6
InChI formel InChI=1S/C27H48/c1-19(2)9-8-10-20(3)23-14-15-24-22-13-12-21-11-6-7-17-26(21,4)25(22)16-18-27(23,24)5/h19-25H,6-18H2,1-5H3/t20-,21-,22+,23-,24+,25+,26+,27-/m1/s1/i6D2,7D2,11D2
SMIL C[C@@]12[C@]([C@]3([C@@]([C@]4(C)[C@@](CC3)(C(C(C(C4)([2H])[2H])([2H])[2H])([2H])[2H])[H])(CC1)[H])[H])(CC[C@@]2([C@@H](CCCC(C)C)C)[H])[H]
IUPAC navn (5R,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-((R)-6-methylheptan-2-yl)hexadecahydro-1H-cyclopenta[a]phenanthrene-2,2,3,3,4,4-d6
Molekylvægt (g/mol) 378.72
Formel vægt 378.41