Få mere at vide
5alpha-Cholestan-3beta-ol-2,2,3,4,4-d5, CDN
Artikelnummer. 30324495
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324495 0.05 g
30317790 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324495 Leverandør CDN Leverandørnr. CDND62560.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5 Alpha-Cholestan-3 Beta-ol-2,2,3,4,4-d5
CAS 2260669-14-3
Molekylær formel C27 D5 H43 O
Mængde 0.05 g
Synonym Cholestan-2,2,3,4,4-d5-3-ol, (3β,5α)- (ACI), (3β,5α)-Cholestan-2,2,3,4,4-d5-3-ol (ACI), 5α-Cholestan-3β-ol-2,2,3,4,4-d5, Cholestanol-d5, Cholestanol-2,2,3,4,4-d5, (3β,5α)-Cholestanol-2,2,3,4,4-d5, Cholestanol-2,2,3,4,4-D5
InChI formel InChI=1 S/C27H48O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h18-25,28 H,6-17H2,1-5H3/t19-,20+,21+,22+,23-,24+,25+,26+,27-/m1/s1/i13D2,17D2,21 D
SMIL [2 H]C1([2 H])C[C@]2(C)[C@H]3 CC[C@]4(C)[C@H](CC[C@H]4 [C@@H]3 CC[C@H]2 C([2 H])([2 H])[C@@]1([2 H])O)[C@H](C)CCCC(C)C
IUPAC navn (3 S,5 S,8 R,9 S,10 S,13 R,14 S,17 R)-2,2,3,4,4-pentadeuterio-10,13-dimethyl-17-[(2 R)-6-methylheptan-2-yl]-1,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-ol
Molekylvægt (g/mol) 393.7002
Formel vægt 393.4019 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity