Få mere at vide
5Alpha-Cholestan-3Alpha-ol-2,2,3,4,4-d5, TRC
Artikelnummer. 30366050
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30366050 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366050 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4317435MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5Alpha-Cholestan-3Beta-ol-2,2,3,4,4-d5
CAS 2260669-14-3
Molekylær formel C27 D5 H43 O
Mængde 5 mg
Synonym Cholestan-2,2,3,4,4-d5-3-ol, (3β,5α)- (ACI), (3β,5α)-Cholestan-2,2,3,4,4-d5-3-ol (ACI), 5α-Cholestan-3β-ol-2,2,3,4,4-d5, Cholestanol-d5, Cholestanol-2,2,3,4,4-d5, (3β,5α)-Cholestanol-2,2,3,4,4-d5, Cholestanol-2,2,3,4,4-D5
InChI formel InChI=1S/C27H48O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h18-25,28H,6-17H2,1-5H3/t19-,20+,21+,22+,23-,24+,25+,26+,27-/m1/s1/i13D2,17D2,21D
SMIL [2H]C1([2H])C[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC[C@H]2C([2H])([2H])[C@@]1([2H])O)[C@H](C)CCCC(C)C
IUPAC navn (3S,5S,8R,9S,10S,13R,14S,17R)-2,2,3,4,4-pentadeuterio-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-ol
Molekylvægt (g/mol) 393.7002
Formel vægt 393.4019