Få mere at vide
5Alpha-Androstane-3Alpha,17Beta-diol-d3, TRC
Artikelnummer. 30330900
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30330900 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30330900 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6375221MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5alpha-Androstane-3alpha,17beta-diol-d3
CAS 79037-33-5
Molekylær formel C19H29D3O2
Mængde 1 mg
Synonym (3α,5α,17β)-Androstane-3,17-diol-d3, 3α,17β-Dihydroxy-5α-androstane-d3, 3α,5α-Tetrahydrotestosterone-d3, Adiol-d3, Androstane-3α,17β-diol-d3, Etiocholane-3α,17β-diol-d3, Hombreol-d3, NSC 9899-d3, 5α-Androstane-3α,17β- diol-16,16,17-d3
InChI formel InChI=1S/C19H32O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12-17,20-21H,3-11H2,1-2H3/t12-,13+,14?,15-,16-,17-,18-,19-/m0/s1/i6D2,17D
SMIL O[C@@]1([2H])C([2H])([2H])C[C@@]2([H])C3CC[C@@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])CC[C@@]21C
IUPAC navn (3R,5S,9S,10S,13S,14S,17S)-16,16,17-trideuterio-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15-dodecahydro-1H-cyclopenta[a]phenanthrene-3,17-diol
Molekylvægt (g/mol) 295.48