Få mere at vide
5alpha-Androstan-3alpha-ol-17-one-16,16-d2, CDN
Artikelnummer. 30325947
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30325947 0.05 g -
30323159 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325947 Leverandør CDN Leverandørnr. CDND54100.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Androsterone-16,16-D2
CAS 89685-22-3
Molekylær formel C19 D2 H28 O2
Mængde 0.05 g
Synonym Androstan-17-one-16,16-d2, 3-hydroxy-, (3α,5α)- (9 CI), 5α-Androstan-3α-ol-17-one-16,16-d2, Androsterone-d2, Androsterone-16,16-d2, Androsterone-16,16-D2, (3α,5α)-3-Hydroxyandrostan-17-one-16,16-d2
InChI formel InChI=1 S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12-16,20 H,3-11H2,1-2H3/t12-,13+,14-,15-,16-,18-,19-/m0/s1/i6D2
SMIL [2 H]C1([2 H])C[C@H]2 [C@@H]3 CC[C@H]4 C[C@H](O)CC[C@]4(C)[C@H]3 CC[C@]2(C)C1=O
IUPAC navn (3 R,5 S,8 R,9 S,10 S,13 S,14 S)-16,16-dideuterio-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15-dodecahydro-1 H-cyclopenta[a]phenanthren-17-one
Molekylvægt (g/mol) 292.4526
Formel vægt 292.2371 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity