Få mere at vide
(5a)-Estrane-3,17-dione-d6, TRC
Artikelnummer. 30379696
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30379696 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379696 Leverandør Toronto Research Chemicals Leverandørnr. TRCE66842225MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (5alpha)-Estrane-3,17-dione-d6
CAS 122605-91-8
Molekylær formel C18H20D6O2
Mængde 25 mg
InChI formel InChI=1S/C18H26O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h11,13-16H,2-10H2,1H3/t11-,13-,14+,15+,16-,18-/m0/s1/i3D2,7D2,10D2
SMIL [2H]C1([2H])C[C@@H]2[C@H]3CC[C@@]4(C)[C@@H](CC([2H])([2H])C4=O)[C@@H]3CC[C@H]2C([2H])([2H])C1=O
IUPAC navn (5S,8R,9R,10S,13S,14S)-2,2,4,4,16,16-hexadeuterio-13-methyl-5,6,7,8,9,10,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthrene-3,17-dione
Molekylvægt (g/mol) 280.43
Formel vægt 280.2309