missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Novus Biologicals COP9 Antibody (2D10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00008533-M02-50ug
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
COP9 Monoclonal antibody specifically detects COP9 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
Tekniske data
| COP9 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| COP9 constitutive photomorphogenic homolog subunit 3 (Arabidopsis), COP9 signalosome complex subunit 3, CSN3COP9 complex subunit 3, JAB1-containing signalosome subunit 3, SGN3COP9 (constitutive photomorphogenic, Arabidopsis, homolog) subunit 3, Signalosome subunit 3 | |
| COPS3 (NP_003644, 324 a.a. ~ 422 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SRVQLSGPQEAEKYVLHMIEDGEIFASINQKDGMVSFHDNPEKYNNPAMLHNIDQEMLKCIELDERLKAMDQEITVNPQFVQKSMGSQEDDSGNKPSSY | |
| 50 μg | |
| Cell Cycle and Replication, Neuronal Cell Markers, Neurotransmission, Signal Transduction | |
| 8533 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA | |
| 2D10 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA | |
| NP_003644 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion