Learn More
Spectrum™ Labs Spectra/Por™ 3 3.5 kD MWCO Standard RC Dry Dialysis Kits
Product Code. 15360772
Change view
Click to view available options
Diameter (Metric):
11.5 mm
29 mm
34 mm
Length (Metric):
1.1 mL/cm vol./length
5 m
6.4 mL/cm vol./length
9.3 mL/cm vol./length
Quantity:
1 m Tubing/Kit
5 m Tubing/Kit
Unit Size:
1m
5m
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) Quantity Unit Size
15360772 29 mm 6.4 mL/cm vol./length 5 m Tubing/Kit 5m
15370772 29 mm 6.4 mL/cm vol./length 1 m Tubing/Kit 1m
15380772 34 mm 9.3 mL/cm vol./length 1 m Tubing/Kit 1m
15340772 11.5 mm 1.1 mL/cm vol./length 1 m Tubing/Kit 1m
12791138 11.5 mm 1.1 mL/cm vol./length 5 m Tubing/Kit Each
11571060 34 mm 5 m 5 m Tubing/Kit 5m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15360772 Supplier Spectrum™ Supplier No. 132724T
  • ONLINE EXCLUSIVE PRICE
    1330.00 DKK
    1162.89 DKK / 5m
    Spare 167.11 DKK
    12% Slukket
Forventet forsendelse: 15-12-2026
Includes
  • 1m or 5m of Spectra/Por 3 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
  • For basic dialysis applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water) or without glycerin (no soaking required)
Specifications
Product Type Dialysis Trial Kit
MWCO 3.5 kDa
Format Tubing
Diameter (Metric) 29 mm
Length (Metric) 6.4 mL/cm vol./length
Quantity 5 m Tubing/Kit