Få mere at vide
Spectrum™ Labs Spectra/Por™ 3 3,5 kD MWCO Standard RC Tørdialysesæt
Diameter (metrisk):
11.5 mm
29 mm
34 mm
Længde (metrisk):
1,1 ml/cm vol./længde
1.1 mL/cm vol./length
5 m
6,4 ml/cm vol./længde
6.4 mL/cm vol./length
9.3 mL/cm vol./length
Mængde:
1 m Tubing/Kit
5 m rør/sæt
Pakningsstørrelse:
1m
5m
Hver
Beskrivelse
Indeholder
- 1m or 5m of Spectra/Por 3 dialysis tubing
- 1 Standard Spectra/Por Closure (for buoyance)
- 1 weighted Spectra/Por Closure (for orientation)
- 2 adhesive tubing openers
- For basic dialysis applications, or evaluating dialysis tubing for dialysis application
- Packaged dry with glycerin (soak in water) or without glycerin (no soaking required)
Tekniske data
Tekniske data
| MWCO | 3.5 kDa |
| Diameter (metrisk) | 11.5 mm |
| Længde (metrisk) | 1.1 mL/cm vol./length |
| Mængde | 1 m Tubing/Kit |