Learn More
Spectrum™ Labs 0.1 - 0.5 kD Biotech CE Dialysis Kits
Product Code. 15330662
Change view
Click to view available options
Diameter (Metric):
6.4 mm
10 mm
15 mm
20 mm
Length (Metric):
0.32 mL/cm vol./length
0.79 mL/cm vol./length
1.8 mL/cm vol./length
3.1 mL/cm vol./length
Unit Size:
1m
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) Unit Size
15330662 15 mm 1.8 mL/cm vol./length 1m
15340662 20 mm 3.1 mL/cm vol./length 1m
15320662 6.4 mm 0.32 mL/cm vol./length 1m
11570940 10 mm 0.79 mL/cm vol./length Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15330662 Supplier Spectrum™ Supplier No. 131057T
  • ONLINE EXCLUSIVE PRICE
    1435.00 DKK
    1268.61 DKK / 1m
    Spare 166.39 DKK
    11% Slukket
Forventet forsendelse: 10-12-2026
Includes
  • 1m Biotech CE Tubing in 0.05% sodium azide
  • Two universal tubing closures
  • Five opening picks
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
  • Hydrophilic, symmetric porositiy
  • For small tubing demand applications or evaluating a specific dialysis membrane
  • Includes 1m tubing with pair of tubing closures and 5 opening picks
Specifications
Product Type Dialysis Tubing Trial Kit
MWCO 0.1 to 0.5 kDa
Format Tubing
Diameter (Metric) 15 mm
Length (Metric) 1.8 mL/cm vol./length
Quantity 1 m Tubing/Kit
Content And Storage Recommended storage in refrigeration (4-8°C).