Learn More
Spectrum™ Labs Spectra/Por™ 7 15,000 D MWCO Standard RC Pre-Treated Dialysis Kits
With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds
1065.90 DKK
Specifications
MWCO | 15 kDa |
---|---|
Quantity | 1 m Tubing/Kit |
Includes:
- 1meter of pre-treated tubing, a pair of closures and 5 opening picks
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
15350702
|
Spectrum™
132 121T |
0.45 mL/cm vol./length | 7.5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15380702
|
Spectrum™
132 124T |
6.4 mL/cm vol./length | 29 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15370702
|
Spectrum™
132 123T |
3.3 mL/cm vol./length | 20.4 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15360702
|
Spectrum™
132 122T |
1.8 mL/cm vol./length | 15 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Hydrophilic, symmetric porositiy
- Equilibrated in water
- Packaged wet in preservative solution of 0.05% sodium azide
- For small experiments or qualifying a membrane for a particular application
Specifications
15 kDa | |
1 m Tubing/Kit |