Learn More
Reagecon 500ML Ion Chromatography Multi Element (3) Anion Cl @150ppm, NO3 @100ppm, SO4 @500ppm in HP Water
BIOPHARMA
Brand:  Reagecon ICA3-MIX2-500
500ML Ion Chromatography Multi Element (3) Anion Cl @150ppm, NO3 @100ppm, SO4 @500ppm in HP Water
Product Code. 16480744
  • 5515.00 DKK / 500mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberICA3MIX222B1 Certificate TypeCertificate of Analysis Date16/04/2022 Catalog NumberICA3-MIX2-500
Promotions
Promotions
Product Content Correction
Product Title
Reagecon 500ML Ion Chromatography Multi Element (3) Anion Cl @150ppm, NO3 @100ppm, SO4 @500ppm in HP Water >