Learn More
Reagecon 500ML Ion Chromatography Multi Element (3) Anion Cl @150ppm, NO3 @100ppm, SO4 @500ppm in HP Water
Brand: Reagecon ICA3-MIX2-500
500ML Ion Chromatography Multi Element (3) Anion Cl @150ppm, NO3 @100ppm, SO4 @500ppm in HP Water
This item is not returnable.
View return policy
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ICA3MIX222B1 | Certificate of Analysis | 16/04/2022 | ICA3-MIX2-500 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Reagecon 500ML Ion Chromatography Multi Element (3) Anion Cl @150ppm, NO3 @100ppm, SO4 @500ppm in HP Water >
Spot an opportunity for improvement?Share a Content Correction