missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Trifluoromethyl-2H-pyridazine-3-one, TRC

Artikelnummer. 30456517
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30456517 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456517 Supplier Toronto Research Chemicals Supplier No. TRCT791350100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 5-Trifluoromethyl-2H-pyridazine-2-one
CAS 244268-34-6
Molekylær formel C5 H3 F3 N2 O
Mængde 100 mg
InChI formel InChI=1S/C5H3F3N2O/c6-5(7,8)3-1-4(11)10-9-2-3/h1-2H,(H,10,11)
SMIL FC(F)(F)C1=CC(=O)NN=C1
IUPAC navn 4-(trifluoromethyl)-1H-pyridazin-6-one
Molekylvægt (g/mol) 164.09
Formel vægt 164.0197
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.