Få mere at vide
MedChemExpress 5-TAMRA-SE
Tekniske data
Tekniske data
| Mængde | 5 mg |
| Produkttype | Dye Reagents |
| Molekylvægt (g/mol) | 527.52 g/mol |
| Opløselighed | DMF : 10 mg/mL (18.96 mM; Need ultrasonic) ∣ DMSO : 5 mg/mL (9.48 mM; Need ultrasonic and warming) |
| Indhold og opbevaring | 4°C, protect from light, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light, stored under nitrogen) |
| Forsendelsestilstand | Room temperature |
| Mål | HBV |
| CAS | 150810-68-7 |
| Synonym | 5-TAMRA-NHS ester ∣ 5-Carboxytetramethylrhodamine succinimidyl ester |