missing translation for 'onlineSavingsMsg'
Learn More

5-Sulfooxymethylfurfural Sodium Salt, TRC

Product Code. 30408814
Change view
Click to view available options
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30408814 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30408814 Supplier Toronto Research Chemicals Supplier No. TRCS68907550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 5-Sulfooxymethylfurfural Sodium Salt
CAS 1330171-28-2
Molekylær formel C6H5NaO6S
Mængde 50 mg
Synonym 5-[(Sulfooxy)methyl]-2-furancarboxaldehyde Sodium Salt, SMF
InChI formel InChI=1S/C6H6O6S.Na/c7-3-5-1-2-6(12-5)4-11-13(8,9)10;/h1-3H,4H2,(H,8,9,10);/q;+1/p-1
SMIL C1=C(OC(=C1)C=O)COS(=O)(=O)[O-].[Na+]
IUPAC navn sodium;(5-formylfuran-2-yl)methyl sulfate
Molekylvægt (g/mol) 228.16
Formel vægt 227.9705
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.