Få mere at vide
5-(Phenylmethoxy)-1H-indole-3-ethan-Alpha,Alpha,Beta,Beta-d4-ol, TRC
Artikelnummer. 30418071
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30418071 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418071 Leverandør Toronto Research Chemicals Leverandørnr. TRCP33639710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-(Phenylmethoxy)-1H-indole-3-ethan-alpha,alpha,beta,beta-d4-ol
CAS 75238-47-0
Molekylær formel C17 D4 H13 N O2
Mængde 10 mg
Synonym 1H-Indole-3-ethan-α,α,β,β-d4-ol','5-(phenylmethoxy)- (9CI), 5-(Phenylmethoxy)-1H-indole-3-ethan-α,α,β,β-d4-ol, 2-(5-Benzyloxy-1H-indol-3-yl)-ethanol, 5-Benzyloxytryptophol-α,α,β,β-d4, 5-Benzyloxy-1H-indole-3-ethanol-D4 (ethanol-D4)
InChI formel InChI=1S/C17H17NO2/c19-9-8-14-11-18-17-7-6-15(10-16(14)17)20-12-13-4-2-1-3-5-13/h1-7,10-11,18-19H,8-9,12H2/i8D2,9D2
SMIL [2H]C([2H])(O)C([2H])([2H])c1c[nH]c2ccc(OCc3ccccc3)cc12
IUPAC navn 1,1,2,2-tetradeuterio-2-(5-phenylmethoxy-1H-indol-3-yl)ethanol
Molekylvægt (g/mol) 271.347
Formel vægt 271.151