Få mere at vide
5-Oxo Pitavastatin, TRC
Artikelnummer. 30423127
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30423127 2.5 mg
30415915 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30423127 Leverandør Toronto Research Chemicals Leverandørnr. TRCO8643502.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 222306-15-2
Molekylær formel C25 H22 F N O4
Molekylvægt (g/mol) 419.44
Synonym (3R,6E)-7-[2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3-hydroxy-5-oxo-6-heptenoic Acid
IUPAC navn (E,3R)-7-[2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl]-3-hydroxy-5-oxohept-6-enoic acid
SMIL O[C@@H](CC(=O)O)CC(=O)\C=C\c1c(nc2ccccc2c1c3ccc(F)cc3)C4CC4
Tekniske data
Kemisk navn eller materiale 5-Oxo Pitavastatin
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C25H22FNO4/c26-17-9-7-15(8-10-17)24-20-3-1-2-4-22(20)27-25(16-5-6-16)21(24)12-11-18(28)13-19(29)14-23(30)31/h1-4,7-12,16,19,29H,5-6,13-14H2,(H,30,31)/b12-11+/t19-/m1/s1
Formel vægt 419.1533
Procent renhed >95%