Få mere at vide
5-Oxo-DL-proline-d5, TRC
Artikelnummer. 30411155
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30411155 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30411155 Leverandør Toronto Research Chemicals Leverandørnr. TRCO8589721MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Oxo-DL-proline-d5
CAS 352431-30-2
Molekylær formel C5 2H5 H2 N O3
Mængde 1 mg
Synonym Proline-2,3,3,4,4-d5','5-oxo- (9CI), 5-Oxoproline-2,3,3,4,4-d5, 5-Oxo-DL-proline-2,3,3,4,4-d5, 2-Pyrrolidone-5-carboxylic-3,3,4,4,5-d5 acid, DL-Pyroglutamic acid-d5, DL-Pidolic Acid-D5, DL-2-Pyrrolidinone-3,3,4,4,5-d5-5-carboxylic Acid, 5-Oxo-DL-proline-d5
InChI formel InChI=1S/C5H7NO3/c7-4-2-1-3(6-4)5(8)9/h3H,1-2H2,(H,6,7)(H,8,9)/i1D2,2D2,3D
SMIL [2H]C1([2H])C(=O)NC([2H])(C(=O)O)C1([2H])[2H]
IUPAC navn 2,3,3,4,4-pentadeuterio-5-oxopyrrolidine-2-carboxylic acid
Molekylvægt (g/mol) 134.14
Formel vægt 134.074