missing translation for 'onlineSavingsMsg'
Få mere at vide

5’-Oxo Amisulpride, TRC

Artikelnummer. 30428779
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428779 5 mg
30416588 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30428779 Leverandør Toronto Research Chemicals Leverandørnr. TRCO8471005MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1391052-47-3
Molekylær formel C17H25N3O5S
Molekylvægt (g/mol) 383.46
Synonym 4-Amino-N-[(1-ethyl-5-oxo-2-pyrrolidinyl)methyl]-5-(ethylsulfonyl)-2-methoxybenzamide
IUPAC navn 4-amino-N-[(1-ethyl-5-oxopyrrolidin-2-yl)methyl]-5-ethylsulfonyl-2-methoxybenzamide
SMIL CCN1C(CNC(=O)c2cc(c(N)cc2OC)S(=O)(=O)CC)CCC1=O

Tekniske data

Kemisk navn eller materiale 5'-Oxo Amisulpride
Mængde 5 mg
InChI formel InChI=1S/C17H25N3O5S/c1-4-20-11(6-7-16(20)21)10-19-17(22)12-8-15(26(23,24)5-2)13(18)9-14(12)25-3/h8-9,11H,4-7,10,18H2,1-3H3,(H,19,22)
Formel vægt 383.1515
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.