missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitropyridine-2-carboxylic acid, ≥97%, Thermo Scientific Chemicals
225.06 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
It plays an important role in synthetic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It plays an important role in synthetic intermediate.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
It plays an important role in synthetic intermediate.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 208°C to 212°C |
Quantity | 1 g |
Beilstein | 139209 |
Solubility Information | Soluble in dimethyl sulfoxide. |
Formula Weight | 168.11 |
Percent Purity | ≥97% |
Physical Form | Solid |
Chemical Name or Material | 5-Nitropyridine-2-carboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'