missing translation for 'onlineSavingsMsg'
Learn More

5-Nitroisoquinoline, 98+%, Thermo Scientific™

Catalog Number 11498186
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
607-32-9
C9H6N2O2
174.159
MFCD00006905
PYGMPFQCCWBTJQ-UHFFFAOYSA-N
isoquinoline, 5-nitro, 5-nitro-isoquinoline, 5-nitro isoquinoline, 5-nitroisochinolin, pubchem6272, maybridge1_001820, acmc-1av1q, 5-nitroisoquinoline, pygmpfqccwbtjq-uhfffaoysa
69085
5-nitroisoquinoline
C1=CC2=C(C=CN=C2)C(=C1)[N+](=O)[O-]
This item is not returnable. View return policy
607-32-9
C9H6N2O2
174.159
MFCD00006905
PYGMPFQCCWBTJQ-UHFFFAOYSA-N
isoquinoline, 5-nitro, 5-nitro-isoquinoline, 5-nitro isoquinoline, 5-nitroisochinolin, pubchem6272, maybridge1_001820, acmc-1av1q, 5-nitroisoquinoline, pygmpfqccwbtjq-uhfffaoysa
69085
5-nitroisoquinoline
C1=CC2=C(C=CN=C2)C(=C1)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 106°C to 110°C
Quantity 5 g
Beilstein 134858
Formula Weight 174.16
Percent Purity ≥98%
Chemical Name or Material 5-Nitroisoquinoline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'