missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Nitro-6-methyluracil, 99%, Thermo Scientific™

Artikelnummer. 15472308
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 g
5 g
Pakningsstørrelse:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
15472308 250 mg 250mg
15452308 1 g 1g
15462308 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15472308 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H55913.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Nitro-6-methyluracil, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in DMF, DMSO, hot methanol and hot water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 16632-21-6
Molekylær formel C5H5N3O4
Molekylvægt (g/mol) 171.11
MDL nummer MFCD00051201
InChI nøgle LIVYMRJSNFHYEN-UHFFFAOYSA-N
Synonym 5-nitro-6-methyluracil, 6-methyl-5-nitrouracil, 6-methyl-5-nitropyrimidine-2,4 1h,3h-dione, 2,4-dihydroxy-6-methyl-5-nitropyrimidine, 6-metyl-5-nitrouracil, 6-methyl-5-nitropyrimidine-2,4-diol, uracil, 6-methyl-5-nitro, 2,4 1h,3h-pyrimidinedione, 6-methyl-5-nitro, 4-methyl-5-nitrouracil, 2,4-dihydroxy-6-methyl-5-*nitropyrimidine
PubChem CID 85523
IUPAC navn 6-methyl-5-nitro-1H-pyrimidine-2,4-dione
SMIL CC1=C(C(=O)NC(=O)N1)[N+]([O-])=O

Tekniske data

Kemisk navn eller materiale 5-Nitro-6-methyluracil
Mængde 250 mg
Opløselighedsinformation Soluble in DMF,DMSO,hot methanol and hot water.
Formel vægt 171.11
Procent renhed 99%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.