missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furaldoxime, 97%, Thermo Scientific™

Catalog Number p-7043578
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
555-15-7
C5H4N2O4
156.10
MFCD00198004,MFCD00198004
PTBKFATYSVLSSD-ZZXKWVIFSA-N
nifuroxime, 5-nitro-2-furaldehyde oxime, usaf ea-5, 2-furaldehyde, 5-nitro-, oxime, 2-furaldoxime, 5-nitro, 5-nitro-2-furaldoxime, carbazylquinone, carbilquinone, 5-nitrofuran-2-carbaldehyde oxime, 5-nitro-2-furancarboxaldehyde oxime
6478035
O\N=C\C1=CC=C(O1)[N+]([O-])=O
This item is not returnable. View return policy
555-15-7
C5H4N2O4
156.10
MFCD00198004,MFCD00198004
PTBKFATYSVLSSD-ZZXKWVIFSA-N
nifuroxime, 5-nitro-2-furaldehyde oxime, usaf ea-5, 2-furaldehyde, 5-nitro-, oxime, 2-furaldoxime, 5-nitro, 5-nitro-2-furaldoxime, carbazylquinone, carbilquinone, 5-nitrofuran-2-carbaldehyde oxime, 5-nitro-2-furancarboxaldehyde oxime
6478035
O\N=C\C1=CC=C(O1)[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 10 g
Beilstein 83037
Sensitivity Light sensitive
Merck Index 14,6536
IUPAC Name (E)-N-[(5-nitrofuran-2-yl)methylidene]hydroxylamine
Formula Weight 156.1
Percent Purity 97%
Chemical Name or Material 5-Nitro-2-furaldoxime
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'