Learn More
Alfa Aesar™ 5-Nitro-2-(4-methylphenyl)benzimidazole, 95%
Brand:  Alfa Aesar™ H64316.06
Additional Details : CAS Number : 1571-90-0 Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15401469
  • 9607.87 DKK / 5g
Estimated Shipment date
from Supplier 17-10-2018
Add to basket
Specifications
Chemical Identifiers
1571-90-0
253.261
AAMPIRQLXWIYCV-UHFFFAOYSA-N
376349
CC1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)[N+](=O)[O-]
C14H11N3O2
MFCD00430513
5-Nitro-2-(p-tolyl)benzimidazole
2-(4-methylphenyl)-6-nitro-1H-benzimidazole
Specifications
5-Nitro-2-(4-methylphenyl)benzimidazole
1571-90-0
C14H11N3O2
5-Nitro-2-(p-tolyl)benzimidazole
CC1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)[N+](=O)[O-]
253.261
253.26
5g
95%
MFCD00430513
AAMPIRQLXWIYCV-UHFFFAOYSA-N
2-(4-methylphenyl)-6-nitro-1H-benzimidazole
376349
Solid