Få mere at vide
5-Nitro-1H-imidazole-1-ethanol-d4, TRC
Artikelnummer. 30470533
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30470533 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470533 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49623210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Nitro-1H-imidazole-1-ethanol-d4
CAS 1794791-19-7
Molekylær formel C5H3D4N3O3
Mængde 10 mg
Synonym 1-(2-Hydroxyethyl)-5-nitroimidazole-d4
InChI formel InChI=1S/C5H7N3O3/c9-2-1-7-4-6-3-5(7)8(10)11/h3-4,9H,1-2H2/i1D2,2D2
SMIL [2H]C([2H])(O)C([2H])([2H])n1cncc1[N+](=O)[O-]
IUPAC navn 1,1,2,2-tetradeuterio-2-(5-nitroimidazol-1-yl)ethanol
Molekylvægt (g/mol) 161.15
Formel vægt 161.0738