Få mere at vide
5-Methyluridine-d4, TRC
Artikelnummer. 30465871
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30465871 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30465871 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3381922.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Methyluridine uridine-d4
CAS 82845-85-0
Molekylær formel C10 2H4 H10 N2 O6
Mængde 2.5 mg
Synonym Uridine-6-d, 5-(methyl-d3)- (9CI), 5-Methyl-d3-uridine-6-d1, 5-Methyl uridine-d4, 5-Methyluridine-d4, 5-Methyluridine-D4 (uridine-D4)
InChI formel InChI=1S/C10H14N2O6/c1-4-2-12(10(17)11-8(4)16)9-7(15)6(14)5(3-13)18-9/h2,5-7,9,13-15H,3H2,1H3,(H,11,16,17)/t5-,6-,7-,9-/m1/s1/i1D3,2D
SMIL [2H]C1=C(C(=O)NC(=O)N1[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C([2H])([2H])[2H]
IUPAC navn 6-deuterio-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(trideuteriomethyl)pyrimidine-2,4-dione
Molekylvægt (g/mol) 262.25
Formel vægt 262.1103