missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-3541135
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
55854-45-0
C5H5ClO2S2
196.66
MFCD05864499
QGGRRHYGHGJEKP-UHFFFAOYSA-N
5-methyl-2-thiophenesulfonyl chloride, 2-thiophenesulfonyl chloride, 5-methyl, 5-methylthiophene-2-sulfonylchloride, 5-methylthiophene-2-sulphonyl chloride, 5-methyl-2-thienylsulfonyl chloride, chloro 5-methyl 2-thienyl sulfone, acmc-209lpu, 5-methylthiophenesulphonyl chloride, 5-methyl-thiophenesulfonyl chloride, 2-chlorosulphonyl-5-methylthiophene
12400112
5-methylthiophene-2-sulfonyl chloride
CC1=CC=C(S1)S(Cl)(=O)=O
This item is not returnable. View return policy
55854-45-0
C5H5ClO2S2
196.66
MFCD05864499
QGGRRHYGHGJEKP-UHFFFAOYSA-N
5-methyl-2-thiophenesulfonyl chloride, 2-thiophenesulfonyl chloride, 5-methyl, 5-methylthiophene-2-sulfonylchloride, 5-methylthiophene-2-sulphonyl chloride, 5-methyl-2-thienylsulfonyl chloride, chloro 5-methyl 2-thienyl sulfone, acmc-209lpu, 5-methylthiophenesulphonyl chloride, 5-methyl-thiophenesulfonyl chloride, 2-chlorosulphonyl-5-methylthiophene
12400112
5-methylthiophene-2-sulfonyl chloride
CC1=CC=C(S1)S(Cl)(=O)=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 196.68
Percent Purity 97%
Chemical Name or Material 5-Methylthiophene-2-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'