missing translation for 'onlineSavingsMsg'
Learn More

5-(Methylthio)thiophene-2-carbaldehyde, 90%, Thermo Scientific™

Catalog Number p-2702390
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
24445-35-0
C6H6OS2
158.233
MFCD00173744
RKXYTFTZODXDEF-UHFFFAOYSA-N
5-methylthio thiophene-2-carbaldehyde, 5-methylthio thiophene-2-carboxaldehyde, 5-methylsulfanyl thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,5-methylthio, 2-formyl-5-methylthiothiphene, 2-formyl-5-methylthio thiophene, 5-methylmercapto-2-thiophenaldehyde, 5-methylthio-2-thiophenecarbaldehyde, 5-methylthiothiophene-2-carbaldehyde, 5-methylmercaptothiophene-2-carbaldehyde
2736997
5-methylsulfanylthiophene-2-carbaldehyde
CSC1=CC=C(S1)C=O
This item is not returnable. View return policy
24445-35-0
C6H6OS2
158.233
MFCD00173744
RKXYTFTZODXDEF-UHFFFAOYSA-N
5-methylthio thiophene-2-carbaldehyde, 5-methylthio thiophene-2-carboxaldehyde, 5-methylsulfanyl thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,5-methylthio, 2-formyl-5-methylthiothiphene, 2-formyl-5-methylthio thiophene, 5-methylmercapto-2-thiophenaldehyde, 5-methylthio-2-thiophenecarbaldehyde, 5-methylthiothiophene-2-carbaldehyde, 5-methylmercaptothiophene-2-carbaldehyde
2736997
5-methylsulfanylthiophene-2-carbaldehyde
CSC1=CC=C(S1)C=O
CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 158.24
Percent Purity 90%
Chemical Name or Material 5-(Methylthio)thiophene-2-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'