missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Methyltetrahydrofolic acid, MedChemExpress

Artikelnummer. 16461027
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16461027 50 mg
16435566 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16461027 Leverandør Medchem Express Leverandørnr. HY113046

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress 5-Methyltetrahydrofolic acid (5-Methyl THF) is a biologically active form of folic acid. 5-Methyltetrahydrofolic acid is a methylated derivate of tetrahydrofolate. 5-Methyltetrahydrofolic acid is the predominant natural dietary folate and the principal form of folate in plasma and cerebrospinal fluid.

5-Methyltetrahydrofolic acid (5-Methyl THF) is a biologically active form of folic acid. 5-Methyltetrahydrofolic acid is a methylated derivate of tetrahydrofolate. 5-Methyltetrahydrofolic acid is the predominant natural dietary folate and the principal form of folate in plasma and cerebrospinal fluid.

Tekniske data

Kemisk navn eller materiale 5-Methyltetrahydrofolic acid
Fysisk form Solid
CAS 134-35-0
Farve Light Yellow
Mængde 50 mg
Renhedsgrad noter Research
Molekylær formel C20H25N7O6
Til brug med (applikation) Metabolism-sugar/lipid metabolism
Synonym 5-Methyl THF
Opløselighedsinformation DMSO : 83.33 mg/mL (181.37 mM; Need ultrasonic)
SMIL O=C(O)CC[C@@H](C(O)=O)NC(C1=CC=C(NCC2N(C)C3=C(NC(N)=NC3=O)NC2)C=C1)=O
Molekylvægt (g/mol) 459.46
Formel vægt 459.46
Procent renhed 99.29%
Grad Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.