missing translation for 'onlineSavingsMsg'
Learn More

5-Methylhexanoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7077675
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
628-46-6
C7H14O2
130.19
MFCD00036494
MHPUGCYGQWGLJL-UHFFFAOYSA-N
isoheptanoic acid, isoenanthic acid, 5-methylcaproic acid, isoamylacetic acid, hexanoic acid, 5-methyl, isovenanthic acid, 5-methylhexylic acid, 5-methyl-hexanoic acid, 5-methyl hexanoic acid, unii-n1ak4x34c2
12344
5-methylhexanoic acid
CC(C)CCCC(O)=O
This item is not returnable. View return policy
628-46-6
C7H14O2
130.19
MFCD00036494
MHPUGCYGQWGLJL-UHFFFAOYSA-N
isoheptanoic acid, isoenanthic acid, 5-methylcaproic acid, isoamylacetic acid, hexanoic acid, 5-methyl, isovenanthic acid, 5-methylhexylic acid, 5-methyl-hexanoic acid, 5-methyl hexanoic acid, unii-n1ak4x34c2
12344
5-methylhexanoic acid
CC(C)CCCC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.9050g/mL
Boiling Point 121.0°C (30.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3CH(CH3)(CH2)3CO2H
Quantity 5 g
Specific Gravity 0.905
Solubility Information Solubility in water: slightly. Other solubilities: soluble in organic solvents
Formula Weight 130.18
Percent Purity 98%
Chemical Name or Material 5-Methylhexanoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methylhexanoic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'