Få mere at vide
5-Methylfuran-3(2H)-one, TRC
Artikelnummer. 30465792
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30465792 250 mg
30475848 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30465792 Leverandør Toronto Research Chemicals Leverandørnr. TRCM236540250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Methylfuran-3(2H)-one
CAS 3511-32-8
Molekylær formel C5 H6 O2
Mængde 250 mg
Synonym 5-Methylfuran-3(2H)-one
InChI formel InChI=1S/C5H6O2/c1-4-2-5(6)3-7-4/h2H,3H2,1H3
SMIL CC1=CC(=O)CO1
IUPAC navn 5-methylfuran-3-one
Molekylvægt (g/mol) 98.1
Formel vægt 98.037